Learn More
Therapak™ Exempt Human Specimen Refrigerated and Frozen Shipping Systems
For diagnostic specimens that must be shipped refrigerated or frozen
Supplier: Therapak™ 56876
Description
- Refrigerated Shipping System is ideal for overnight shipments via common carriers; includes small insulated cooler; transport box; 8 oz. cold packs; specimen bag; absorbent material; Exempt Human Specimen and biohazard labels; and cushioning material
- Frozen Shipping System designed for overnight shipments of frozen (dry ice) specimens by common carriers; includes small insulated cooler; transport box; specimen bag; absorbent material; Exempt Human Specimen, biohazard, and dry ice labels, and cushioning material
- Combination Shipping System for shipments of frozen and ambient specimens; includes small insulated cooler; insulated envelope; two biohazard bags; two Aqui-Pak™ pouches (each holds four tubes); ambient gel wrap; transport box; Exempt Human Specimen, biohazard, and dry ice labels, and cushioning material
Specifications
Includes small insulated cooler; transport box, 8 oz. cold packs, specimen bag, absorbent material; EHS and biohazard labels; Cushioning material | |
For overnight shipping |