Learn More
Fisherbrand™ Thermal Mailer with Tape and Zipper Closure
Fisherbrand Thermal Mailer with Tape and Zipper Closure is ideal for short trips and whenever maintaining the cold chain of pharmaceuticals or laboratory products as required.
 View more versions of this product
Catalog No. 07-100-105
Add to cart
Description
Description
Features:
  • Thick padded mailers cushion and insulate up to four hours
  • Dual closure with leakproof zipper inside and exterior self-seal strip
  • Very thick foam (0.5in.) with metallized poly exterior
  • Made in USA
Order Info
Case quantity only 25 per case
Warranty and Services
Warranty against defects in material or workmanship.
Specifications
Specifications
White Tape, Dual closure with leakproof zipper inside and exterior self-seal strip
Outside: 12in x 16in; Inside: 11in x 14in
305mm
25
406mm
Metalized PET Silver Exterior
24.5mm
Outer Layer: Metalized PET laminated to PE, Middle Layer: Open Cell PU Foam, Inner Layer: White LDPE (FDA approved for incidental food contact).
1/2in thickness
Thermal Mailer