Globe Scientific Thermal Printer Paper for Clinical Chemistry Analyzers

Thermal printer paper for use with popular chemistry analyzers.

Manufacturer: Globe Scientific 7146

 View more versions of this product

Catalog No. 21-200-106

  Check Availability
Add to Cart

    Specifications

    For Use With (Equipment) Sysmex CA 50, 500, 1000, 1500 and 5000 analyzers
    Packaging Quantity 5 rolls/box; 48 rolls/case
    Product Type Printer Paper
    Description 58mm wide x 45mm dia. x 80 ft. long

    • Size (L x W): 80 ft. x 58mm
    • Dia.: 45mm
    • Quantity: 5 rolls/box; 48 rolls/case

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit