Learn More
Thermo Scientific 0.2 mL Individual Tubes
0.2 mL individual tubes with attached flat or dome snap shut caps, available in a variety of colors
Specifications
Volume (Metric) | 0.2 mL |
---|---|
Certifications/Compliance | RNase, Dnase and human DNA free |
Type | Thin Wall PCR Tube |
Format | Tube |
For Use With (Equipment) | 0.2mL thermal cycler blocks for PCR and sample storage applications |
Catalog Number | Mfr. No. | Closure Type | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Closure Type | Color | Price | Quantity & Availability | ||||
AB-0337G
|
Thermo Scientific
AB0337G |
Domed | Green |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. Part Not Found |
N/A |
AB-0337B
|
Thermo Scientific
AB0337B |
Domed | Blue |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. Part Not Found |
N/A |
AB-0337R
|
Thermo Scientific
AB0337R |
Domed | Red |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. Part Not Found |
N/A |
AB-0337P
|
Thermo Scientific
AB0337P |
Domed | Purple |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. Part Not Found |
N/A |
AB-0337Y
|
Thermo Scientific
AB0337Y |
Domed | Yellow |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. Part Not Found |
N/A |
AB-0491
|
Thermo Scientific
AB0491 |
Domed Frosted | Assorted |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. Part Not Found |
N/A |
Description
0.2 mL Individual Tubes with integral “snap shut” caps are suitable for 0.2 mL thermal cycler blocks. They have a 0.25 mL maximum volume when closed. These individual tubes feature thin-wall design and are made from virgin polypropylene. The tubes are cleanroom produced, ensuring that they are free from contaminants that could affect experimental results. They are available in multiple colors with flat or domed caps.
The Thermo Scientific 0.2 mL Individual Tubes with integral ”snap shut“ caps are suitable for 0.2 mL thermal cycler blocks. They have a 0.25 mL maximum volume when closed. These individual tubes feature thin-wall design and are made from virgin polypropylene. The tubes are cleanroom produced, ensuring that they are free from contaminants that could affect experimental results. They are available in multiple colors with flat or domed caps.
Highlights
· Suitable for 0.2 mL thermal cycler blocks
· 0.25 mL maximum tube capacity when closed
·
Integral 'snap shut' cap
· Available with flat or domed caps
Applications
· PCR
· Sample storage
Hazardous:No
>Storage Condition:Ambient
PCR; Sample storage
Certifications
Certified free from human DNA, RNases, and DNases
Specifications
0.2 mL | |
Thin Wall PCR Tube | |
0.2mL thermal cycler blocks for PCR and sample storage applications | |
1000 Case |
RNase, Dnase and human DNA free | |
Tube | |
Polypropylene |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.