Thermo Scientific™ 1300 Series Class II, Type A2 Biological Safety Cabinet Packages
Designed for working with biological agents at Biosafety Levels 1–3
Thermo Scientific 1300 Series A2 features advancements in brushless DC motor technology for dramatically improved energy efficiency, safety performance, and reliability. SmartClean offers a window cleaning position that provides the user access to interior of window sash. SmartPort provides a clean and easy way to organize the interior of the 1300 Series A2, with confidence due to our negative pressure dual-side wall.
$9155.00 - $12242.00
Specifications
Filter Efficiency 99.995% MPPS
Metric Centralized Load Capacity 50kg
English Centralized Load Capacity 110 lb.
Certifications NSF/ANSI 49, UL, CE
Voltage 120V
View More Specs
  • Products 16
  • Description & Specifications
Products
Catalog Number Mfr. No. Metric Sash Height English Sash Height English Width Metric Width Price Quantity & Availability    
13-261-340
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
1340
20.3cm 8 in. 3 ft. 91.4cm Each for $9,155.00
Check Availability
Add to Cart
13-261-342
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
1342
20.3cm 8 in. 3 ft. 91.4cm Each for $9,602.00
Check Availability
Add to Cart
13-261-332
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
1321
25cm 10 in. 3 ft. 91.4cm Each for $9,155.00
Check Availability
Add to Cart
13-261-334
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
1323
25.4cm 10 in. 3 ft. 91.4cm Each for $9,602.00
Check Availability
Add to Cart
13-261-314
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
1395
20.3cm 8 in. 4 ft. 122cm Each for $9,715.00
Check Availability
Add to Cart
13-261-312
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
1335
20.3cm 8 in. 4 ft. 122cm Each for $10,161.00
Check Availability
Add to Cart
13-261-304
Available on GSA/VA Contract for Federal Government customers only. 
THERMO SCI ENCLOSURES
1385
25.4cm 10 in. 4 ft. 122cm Each for $9,715.00
Check Availability
Add to Cart
13-261-222
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
1375
25.4cm 10 in. 4 ft. 122cm Each for $10,161.00
Check Availability
Add to Cart
13-261-344
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
1351
20.3cm 8 in. 5 ft. 152.4cm Each for $11,101.00
Check Availability
Add to Cart
13-261-346
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
1353
20.3cm 8 in. 5 ft. 152.4cm Each for $11,572.00
Check Availability
Add to Cart
13-261-336
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
1369
20.3cm 10 in. 5 ft. 152.4cm Each for $11,101.00
Check Availability
Add to Cart
13-261-338
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
1371
20.3cm 10 in. 5 ft. 152.4cm Each for $11,572.00
Check Availability
Add to Cart
13-261-315
Thermo Scientific
1397
Each for $11,774.00
Check Availability
Add to Cart
13-261-313
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
1337
20.3cm 8 in. 6 ft. 183cm Each for $12,242.00
Check Availability
Add to Cart
13-261-306
Available on GSA/VA Contract for Federal Government customers only. 
THERMO SCI ENCLOSURES
1387
25.4cm 10 in. 6 ft. 183cm Each for $11,774.00
Check Availability
Add to Cart
13-261-223
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
1377
25.4cm 10 in. 6 ft. 183cm Each for $12,242.00
Check Availability
Add to Cart
Descriptions & Specifications
Related Products