Learn More
Thermo Scientific™ 384-Well Microtiter Microplates
Excellent for a number of applications.
Manufacturer:  Thermo Scientific™ 95040330
 View more versions of this product
Catalog No. 21-377-345
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Yes
Black
Polystyrene
384-Well Microtiter Microplate
Round
Yes
For high throughput screening, PCR, mutagenicity/mutational spectra assays
Yes
0.07mL