Learn More
Thermo Scientific 4-Place Swinging Bucket Rotor Adapters
For added processing versatility
$88.00 - $92.00
Catalog Number | Mfr. No. | Tube Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Tube Type | Price | Quantity & Availability | |||||
75-007-544
![]() |
Thermo Scientific
75007544 |
3 x 15 mL Blood Tubes |
Each for $88.00
|
|
75-007-557
![]() |
Thermo Scientific
75007557 |
Each for $91.00
|
|
75-007-556
![]() |
Thermo Scientific
75007556 |
1 x 50 mL Falcon Tube |
Each for $92.00
|
|
75-007-546
|
Thermo Scientific
75007546 |
3 x 7 mL Blood Tubes |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
For added processing versatility
- Maximum tube size: 30 x 117mm
Order Info
Order 4-Place Swinging Bucket Rotor (75-007-591) with Round Buckets (75-007-555). Adapters are sold individually; order quantity to equip rotor. Note: 50mL conical adapter (75-007-556) requires rubber pads (supplied with buckets). Remove pads when using all other size adapters.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Swinging bucket rotor |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.