Learn More
Thermo Scientific™ ABgene™ 96-Well PCR CyclePlate
Universal fit in all leading thermal cyclers and MegaBACE™ capillary sequencers
Manufacturer:  Thermo Scientific™ 1055000
Catalog No. 1055000
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • 96-well PCR plate
  • 340°l maximum well volume (when used with adhesive and heat seals)
  • Clear well bottom for sample visibility
  • Highly rigid plate deck
  • Cut-away corner
  • Sealable using Thermo-Mats™, Adhesive Films and Foils, Heat Seals and Flat and Domed Cap Strips
Specifications
Specifications
Standard
PCR
96
Natural
10/Cs.
8 x 12