Learn More
Thermo Scientific™ Accessories for RadEye™ Radiation Detectors
Accessory for use with RadEye pocket-sized radiation detectors
Supplier: Thermo Scientific™ 425067065
Description
- Docking Station/Car Adapter includes charging circuitry (8-30 V DC, cigarette lighter plug), alarm relay, RS 232 interface, AAA NiMH rechargeable batteries
- Goose neck mounting kit has suction cup for instanteneous attachment to windshield; fits to docking station (19-172-509) or desktop holder
- Headset for RadEye includes earphone for RadEye series
Specifications
Docking Station |