Thermo Scientific™ Accessory Rotors for Centra-CL2 Centrifuges Four-Place Swinging Bucket Rotor Packages Available on GSA/VA Contract for Federal Government customers only.
Manufacturer: Thermo Scientific™ 523680F
 View more versions of this product
Catalog No. 05-101-11
  Check Availability
Add to Cart
    Specifications
    Type Rotor
    No. of Positions 4.0
    Capacity (Metric) 5 to 10mL
    Includes Four inserts for 7 to 10mL Blood tubes and four inserts for 5 to 7mL Blood tubes
  • Description & Specifications
Related Products
Product Content Feedback