Learn More
Thermo Scientific™ ALPS 25 Manual Heat Sealer
Protect contents in storage microplates with this simple-to-use, hand-operated sealer designed for low-throughput labs.
Manufacturer:  Thermo Scientific™ AB0384/240
 View more versions of this product
Catalog No. 14-387-304
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
Protect contents in storage microplates with the Thermo Scientific™ ALPS 25 Manual Heat Sealer, a simple-to-use, hand-operated sealer designed for low-throughput labs. Heat sealing offers superior sample protection of storage and reaction plates in compound storage, sample archiving and PCR applications.
Sealing Flexibility:
  • Compatible with virtually any height microplate or block, including PCR plates
  • Choose from a wide range of heat seals for any application, including piercable, optically clear and permanent
Safe, Compact Design:
  • Built-in thermostat that controls trigger automatic system shutoff to prevent overheating of the ALPS 25 sealer
  • Lightweight with small footprint for ease of placement and relocation
Compatible with:
The ALPS 25 sealer is part of a complete line of microplate heat sealing instruments. For enhanced ergonomics and variable time and temperature settings, consider the semi-automated ALPS 50 V microplate heat sealer. For automated microplate sealing, see the ALPS 3000.
Description & Specifications
Specifications
7.5 lb.
10 to 35°C
3.4kg
240V
1
220°C
Manual Heat Sealer
10 to 80%
CE marked
1