Learn More
Thermo Scientific™ Aquasil™ C18 Javelin™ Guard Columns
Offer convenient, easy-to-use protection for analytical HPLC columns
Supplier:  Thermo Scientific™ 77503012106
 View more versions of this product
Catalog No. 77503012106
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • Direct-connection, finger-tight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
Features of Aquasil C18 Solid Phase
  • Thermo Scientific polar endcapped C18 phase
  • Alternative selectively compared to conventional C18 phases
  • Retention of polar compounds
  • Compatible with 100% aqueous mobile phases
TRUSTED_SUSTAINABILITY
Specifications
Specifications
3 μm
10 mm
2.1 mm
C18
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Accessories