Thermo Scientific™ ART™ Barrier Pipette Tips with Breakthru Economy Packaging Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: Thermo Scientific™ 355150

 View more versions of this product

Catalog No. 21-403-05

  Check Availability
Add to Cart

    Specifications

    Product Type Tip
    For Use With (Equipment) Research grade pipetters
    Sterility Sterile
    Description ART PURE 200
    Tip Style ART Barrier, MicroPoint
    Certifications/Compliance Certified Pure
    Packaging Rack
    Volume (Metric) 10 to 200μL
    Color Clear

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit