Thermo Scientific™ ART™ Non-Filtered Gel 10 Eppendorf-Style pipette Tips
Eppendorf™-style pipet designed for loading specific types of gels
Manufacturer: Thermo Scientific™ 3652
 View more versions of this product
Catalog No. 21-403-602
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Product Type Tip
    Material Polyacrylamide
    Length (English) 2.25 in.
    Sterility Sterile
    Tip Style Gel, Round
    Shape Ultra Micro
    Quantity 1020 Pack
    Thickness 0.4mm
    For Use With (Application) For loading specific types of gels
    Volume (Metric) 10μL
  • Description & Specifications
Related Products
Product Content Feedback