Learn More
Thermo Scientific™ ART™ Non-Filtered Low Retention Pipette Tips Available on GSA/VA Contract for Federal Government customers only.
Specifically designed for eliminating sample retention within the pipette tip
Manufacturer:  Thermo Scientific™ 373205HR
 View more versions of this product
Catalog No. 21-402-576
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
10 to 300μL
Clear
Hinged Rack, 10 x 96
Nonsterile
Low Retention
Rainin LTS Pipetters