Thermo Scientific™ ART™ Non-Filtered Low Retention Pipette Tips Available on GSA/VA Contract for Federal Government customers only.
Specifically designed for eliminating sample retention within the pipette tip
Manufacturer: Thermo Scientific™ 375105HR
 View more versions of this product
Catalog No. 21-402-564
  Check Availability
Add to Cart
    Specifications
    Packaging Hinged Rack, 10 x 96
    For Use With (Equipment) Rainin LTS Pipetters
    Sterility Sterile
    Tip Style Low Retention
    Volume (Metric) 0.5 to 250μL
  • Description & Specifications
Related Products
Product Content Feedback