Thermo Scientific™ ART™ Non-Filtered Pipet Tips with MicroPoint™ Tip Design
MicroPoint design means the tip profile is nearly 50% smaller than standard blunt-end tips and significantly more narrow than beveled-end tips
Manufacturer: Thermo Scientific™ 379111
 View more versions of this product
Catalog No. 21-403-09
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Volume (Metric) 100 to 1000μL
    Sterility Sterile
    Color Clear
    Model Pure 1000 REACH
    Tip Style SoftFit Design
    Material Polypropylene
    Packaging Rack
    Length (English) 5 in.
    • Same superior quality and performance as all Thermo Scientific ART pipet tips
    • Crystal clear tips allow you to easily see transparent samples
    • Reference marks on many Thermo Scientific ART tips provide a quick and convenient guide to visually establish whether you are pipetting the desired volume of liquid
    • ART 10 tips are ideal for low volume samples
    • The extended length ART 10 REACH™ and ART 1000 REACH prevent contamination of samples, tubes, and pipetters
    • Ultra Micro and Pure Ultra Micro are also well suited for work with low-volume samples
    • ART 200 are equivalent to the “Universal Yellow“ tip and have a reference mark at 10µL
    ART Non-Filtered tips are available in yellow and blue colors, graduated, beveled tips and large volumes.
  • Description & Specifications
Related Products
Product Content Feedback