Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Barnstead™ Economy Filters and Holders

Supplier:  Thermo Scientific™ D0748

 View more versions of this product

Catalog No. 09-034-48


Add to Cart

Description

Description

Prefilters

  • Remove suspended particles from feedwater
  • Contain cellulose acetate fibers wound in precise patterns on polypropylene core
  • Overall: 10 × 2.5 in. (25 × 6.3cm)
Submicron Postfilters
  • Remove all particulates and microorganisms larger than rated pore size
  • 10 in. (25cm) double open-ended cartridges have 2 Buna-N gaskets for positive seal
  • Pleated membrane has 4.75 sq. ft. (0.46m2) surface area
  • Recommended flowrate: 2 gal./min. (7.5L/min.)
  • Autoclavable at 118°C (245°F)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

2 gal./min. (7.5 L/min.)
0.45 μm
Economy filter holders 09-034-155, 09-034-156
Economy Filter and Holder
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit