Thermo Scientific™ Barnstead™ ROpure LP™ Membranes
Manufacturer: Thermo Scientific™ D2730
 View more versions of this product
Catalog No. 09-034-177B
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Flow Rate 0.25
    Feedwater Temperature (English) 77°F
    Feedwater Temperature (Metric) 25°C
    Product Type Cellulose Acetate membrane
    For Use With (Equipment) RoPure LP system
    • Flowrates in table are for new membranes under these conditions:
        25°C (77°F) feedwater temperature
      • Feedwater composition of 1000ppm NaCl at a pH of 6.0 to 6.5
      • Stated design specifications for operating pressure and feedwater recovery
    • Membrane life expectancy will vary with feedwater conditions from one to three years
  • Description & Specifications
Related Products
Product Content Feedback