Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific BetaBasic 18 DASH-HTS HPLC Columns, 5µm Particle Size
Designed for high throughput, these short columns are excellent for ballistic gradients, with retention and sensitivity for very fast LC/MS applications
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
71505012150
|
Thermo Scientific
71505012150 |
N/A | Particle Size: 5µm; 10L x 2.1mm I.D.; 1/pk. |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
71505022150
|
Thermo Scientific
71505022150 |
N/A | Particle Size: 5µm; 20L x 2.1mm I.D.; 1/pk. |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
- Patented, low dead volume design minimizes band spreading
- Finger-tight fittings make changing columns quick and easy
Features of BetaBasic 18 Solid Phase
- Highly efficient and reproducible columns
- Pore size suitable for small molecules, peptides and protein digests
- Superb pH stability
Specifications
C18 | |
2.1 mm | |
5 μm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title