Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ BetaBasic 4 CPF HPLC Columns
A good first choice for compounds with poor peak symmetry on other columns
Specifications
Particle Size | 5 μm |
---|---|
Pore Size | 150 Å |
USP Type | L26 |
Stationary Phase | C4 |
Packing Material | Silica |
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
71605151032
|
Thermo Scientific™
71605151032 |
Particle Size: 5μm; 150 x 1.0mm I.D. | Particle Size: 5μm; 150 x 1.0mm I.D. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
71605152132
|
Thermo Scientific™
71605152132 |
Particle Size: 5μm; 150 x 2.1mm I.D. | Particle Size: 5μm; 150 x 2.1mm I.D. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
71605153032
|
Thermo Scientific™
71605153032 |
Particle Size: 5μm; 150 x 3.0mm I.D. | Particle Size: 5μm; 150 x 3.0mm I.D. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
71605154632
|
Thermo Scientific™
71605154632 |
Particle Size: 5μm; 150 x 4.6mm I.D. | Particle Size: 5μm; 150 x 4.6mm I.D. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
71605251032
|
Thermo Scientific™
71605251032 |
Particle Size: 5μm; 250 x 1.0mm I.D. | Particle Size: 5μm; 250 x 1.0mm I.D. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
71605252132
|
Thermo Scientific™
71605252132 |
Particle Size: 5μm; 250 x 2.1mm I.D. | Particle Size: 5μm; 250 x 2.1mm I.D. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
71605253032
|
Thermo Scientific™
71605253032 |
Particle Size: 5μm; 250 x 3.0mm I.D. | Particle Size: 5μm; 250 x 3.0mm I.D. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
Thermo Scientific HPLC columns are manufactured under strict ISO 9001:2000 procedures, and every batch of silica and bonded phase is thoroughly tested for physical and chromatographic characteristics. Test reports are included with every column shipped.
- With Integral Column Plus Filter (CPF) System
- Fully deactivated Type B silica to ensure sharp peaks and excellent reproducibility
- Give less retention than C18 or C8 phases, and are a good choice for fast analyses including LC/MS of all types of compounds
- For basic compounds at high pH, which allows for a wide choice of buffer selection in LC/MS applications
- Stable bonding chemistry and excellent surface coverage gives the columns exceptional low and high pH stability
Specifications
5 μm | |
L26 | |
Silica | |
Reversed Phase | |
Reversed Phase |
150 Å | |
C4 | |
6% | |
Yes |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title