Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BetaBasic™ 4 DASH-HTS HPLC Columns

Designed for high throughput, these short columns are excellent for ballistic gradients, with retention and sensitivity for very fast LC/MS applications

Specifications

Stationary Phase C4
Diameter (Metric) 2.1 mm
Length (Metric) 20 mm
Particle Size 5 μm
View More Specs

Products 2
Catalog Number Mfr. No. Particle Size Length (Metric) Diameter (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Particle Size Length (Metric) Diameter (Metric) Price Quantity & Availability  
71605022150
View Documents
Thermo Scientific™
71605022150
5 μm 20 mm 2.1 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
71605022151
View Documents
Thermo Scientific™
71605022151
5 μm 20 mm 2.1 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Thermo Scientific HPLC columns are manufactured under strict ISO 9001:2000 procedures, and every batch of silica and bonded phase is thoroughly tested for physical and chromatographic characteristics. Test reports are included with every column shipped.

  • Fully deactivated Type B silica to ensure sharp peaks and excellent reproducibility
  • Give less retention than C18 or C8 phases, and are a good choice for fast analyses including LC/MS of all types of compounds
  • For basic compounds at high pH, which allows for a wide choice of buffer selection in LC/MS applications
  • Stable bonding chemistry and excellent surface coverage gives the columns exceptional low and high pH stability
  • Patented, low dead volume design minimizes band spreading
  • Finger-tight fittings make changing columns quick and easy
  • Available in packs of 3 or packs of 10 for better cost-effectiveness

Specifications

Specifications

C4
2.1 mm
20 mm
5 μm
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit