Learn More
Thermo Scientific™ BetaBasic™ 4 Guard Cartridges
The Thermo Scientific BetaBasic 4 Guard Cartridges offer convenience, economy and effective protection for extending column lifetime.
Specifications
Length (Metric) 10 mm
Stationary Phase BetaBasic 4
View More Specs
Products 4
Catalog Number Mfr. No. Diameter (Metric) Inner Particle Size Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Inner Particle Size Price Quantity & Availability  
71603012101
Accessories View Documents
Thermo Scientific™
71603012101
2.1 mm 3 µm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
71603014001
Accessories View Documents
Thermo Scientific™
71603014001
4 mm 3 µm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
71605013001
Accessories View Documents
Thermo Scientific™
71605013001
3 mm 5 µm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
71605014001
Accessories View Documents
Thermo Scientific™
71605014001
4 mm 5 µm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • The 10mm design offers maximum protection with minimal increase in retention
  • Fit Thermo Scientific UNIGUARD™ direct connection and stand alone holders
  • Highly efficient and reproducible Thermo Scientific columns
  • 150Å pore size for small molecules, peptides and protein digests
  • Superb pH stability
Specifications
Specifications
10 mm
BetaBasic 4