Learn More
Thermo Scientific™ BetaBasic™ 4 HPLC Columns, 3µm Particle Size Available on GSA/VA Contract for Federal Government customers only.
A good first choice for compounds with poor peak symmetry on other columns
Manufacturer:  Thermo Scientific™ 71603052130
 View more versions of this product
Catalog No. 71603052130
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
2.1mm
50mm
3μm
BetaBasic™ 4