Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BetaBasic™ 8 Columns, Particle size 3μm

Highly efficient and reproducible

Specifications

Particle Size 3 μm
pH 2 to 9
Pore Size 150 Å
Surface Area 200 m²/g
USP Type L7
View More Specs

Products 3
Catalog Number Mfr. No. Diameter (Metric) Length (Metric) Stationary Phase Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Length (Metric) Stationary Phase Price Quantity & Availability  
71403022130
View Documents
Thermo Scientific™
71403022130
2.1 mm 20 mm C8 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
71403053030
View Documents
Thermo Scientific™
71403053030
3 mm 50 mm C8 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
71403153030
View Documents
Thermo Scientific™
71403153030
3 mm 150 mm C8 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Pore size suitable for small molecules, peptides and protein digests
  • Superb pH stability

Order Info

Other column dimensions are also available. Please call Customer Service for more information.

Specifications

Specifications

3 μm
150 Å
L7
Silica
Reversed Phase
Reversed Phase
2 to 9
200 m²/g
C8
7%
Yes
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit