Learn More
Thermo Scientific™ BetaBasic™ CN Javelin™ Guard Columns
Offer convenient, easy to use protection for analytical HPLC columns
Specifications
Particle Size 5 µm
Stationary Phase CN
View More Specs
Products 3
Catalog Number Mfr. No. Diameter (Metric) Inner Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Inner Length (Metric) Price Quantity & Availability  
71705012106
View Documents
Thermo Scientific™
71705012106
2.1 mm 10 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
71705022106
View Documents
Thermo Scientific™
71705022106
2.1 mm 20 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
71705024006
View Documents
Thermo Scientific™
71705024006
4 mm 20 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
Specifications
Specifications
5 µm
CN