Learn More
Thermo Scientific™ BetaBasic™ CN Javelin™ Guard Columns
Offer convenient, easy to use protection for analytical HPLC columns
Supplier:  Thermo Scientific™ 71705012106
 View more versions of this product
Catalog No. 71705012106
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5 μm
10 mm
2.1 mm
CN
Product Suggestions
Product Suggestions