Learn More
Thermo Scientific™ BetaBasic Phenyl Guard Cartridges Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Thermo Scientific™ 71803011001
 View more versions of this product
Catalog No. 71803011001
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
BetaBasic™ HPLC column
Yes
3μm
10mm
Phenyl
7%
Spherical
150Å
3mm