Learn More
Thermo Scientific BetaSil C18 Guard Cartridges
Economically extends HPLC column lifetime
$512.50 - $534.00
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Particle Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Particle Size | Price | Quantity & Availability | ||||
70103012101
|
Thermo Scientific
70103012101 |
2.1 mm | 3 µm |
Pack of 4 for $534.00
|
|
70105013001
![]() |
Thermo Scientific
70105013001 |
3 mm | 5 µm |
Pack of 4 for $512.50
|
|
70105014001
|
Thermo Scientific
70105014001 |
4 or 4.6 mm | 5 µm |
Pack of 4 for $512.50
|
|
70105012101
![]() |
Thermo Scientific
70105012101 |
2.1 mm | 5 µm |
Pack of 4 for $512.50
|
|
70105011001
|
Thermo Scientific
70105011001 |
1 mm | 5 µm |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
70103011001
|
Thermo Scientific
70103011001 |
1 mm | 3 µm |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
70103013001
|
Thermo Scientific
70103013001 |
3 mm | 3 µm |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
70103014001
|
Thermo Scientific
70103014001 |
4 mm | 3 µm |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
Features of Betasil C18 Solid Phase
- Thermo Scientific Chemistry-on 100&Angst; silica
- High surface area with high bonded phase coverage
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
10 mm | |
C18 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.