Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BetaSil™ Phenyl Guard Cartridges

Drop-in guard cartridges offer convenience, economy and effective protection for extending column lifetime

Supplier:  Thermo Scientific™ 70605011001

 View more versions of this product

Catalog No. 70605011001

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Requires : Thermo Scientific Uniguard™ or standalone holder, sold separately.

Description

Description

  • Thermo Scientific chemistry on 100Å silica
  • High surface area with high bonded phase coverage
  • Particle size: 5μm
  • 10mm x 4.0mm I.D.
  • The 10mm design offers maximum protection with minimal increase in retention
  • Fit Thermo Scientific UNIGUARD™ direct connection and stand alone holders

TRUSTED_SUSTAINABILITY
Specifications

Specifications

5 μm
10 mm
1 mm
Phenyl
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Accessories
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit