Learn More
Thermo Scientific™ BioBasic™ 4 Javelin™ Guard Columns
Javelin guard columns offer convenient, easy to use protection for analytical HPLC columns
$564.00
Specifications
Stationary Phase BioBasic 4
Particle Size 5 µm
View More Specs
Products 1
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Inner Price Quantity & Availability  
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Inner Price Quantity & Availability  
72305021006
View Documents
Thermo Scientific™
72305021006
20 mm 1 mm
Pack of 4 for $564.00
Add to cart
Description
Description
  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
  • Based on 300Å silica for outstanding biomolecule performance
  • Lower carbon load for optimal retention of larger peptides and proteins
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations
Specifications
Specifications
BioBasic 4
5 µm