Learn More
Thermo Scientific™ BioBasic™ 4 Javelin™ Guard Columns Available on GSA/VA Contract for Federal Government customers only.
Javelin guard columns offer convenient, easy to use protection for analytical HPLC columns
Manufacturer:  Thermo Scientific™ 72305021006
 View more versions of this product
Catalog No. 72305021006
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
5μm
20mm
BioBasic 4
1mm