Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BioBasic™ SEC 60A HPLC Columns

Outstanding size separation of water soluble compounds

Specifications

Column Format Analytical Column
Length (Metric) 150 mm
Particle Size 5 μm
Pore Size 60 Å
Stationary Phase Proprietary
View More Specs

Products 1
Catalog Number Mfr. No. Diameter (Metric) Length (Metric) Stationary Phase Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Length (Metric) Stationary Phase Price Quantity & Availability  
73305154630
View Documents
Thermo Scientific™
73305154630
4.6 mm 150 mm Proprietary This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Covers separation of analytes over a wide molecular weight range
  • Long column life and high column efficiencies
  • Simple mechanism of interaction based on molecular size and shape
  • Excellent for sample cleanup
  • Straghtforward method development, simple mobile phases
  • 60Å pore size
  • Particle size: 5μm

Order Info

For information on bulk quantities, please inquire.

Specifications

Specifications

Analytical Column
5 μm
Proprietary
Silica
Size Exclusion and Gel-Permeation
150 mm
60 Å
BioBasic
Mixed Mode
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit