Learn More
Fisherbrand™ Castaloy™ Hosecock Clamp
Fine-machined adjustment screw for very accurate regulation of flow
$26.75
Specifications
Type Hosecock Clamp
For Use With (Application) For accurate regulation of flow
Size (English) Grip 0.75 in. O.D.
Size (Metric) Grip 19 mm O.D.
View More Specs
Products 1
Catalog Number Includes Price Quantity & Availability  
Catalog Number Includes Price Quantity & Availability  
05-847Q
View Documents
Adjustment screw
Each for $26.75
Add to cart
Description
Description
Fine-machined adjustment screw for very accurate regulation of flow
  • Threaded keeper screw on side of clamp can be raised to allow easy positioning of tubing, then screwed back into base of clamp
  • For tubing up to 0.75 in. (19mm) O.D.
  • Concave lower bearing surface, convex upper which reduces amount of pressure required for complete closure
Specifications
Specifications
Hosecock Clamp
For accurate regulation of flow
0.75 in. O.D.
19 mm O.D.