Fisherbrand™ Castaloy™ Regular or Heavy-Duty Pinchcocks
Complete closure
 View more versions of this product
Catalog No. 05-849BQ
  Check Availability
Add to Cart
    Specifications
    Product Type Heavy Duty Pinchcock
    For Use With (Equipment) Works well with stiff tubing, used in gas and water connections
    For Use With (Application) To compress a flexible tube to control the flow of fluid through it
  • Description & Specifications
Related Products
Product Content Feedback