Learn More
Fisherbrand™ Castaloy™ Two-Prong Extension Clamps
Flat, vinyl- or fiberglass-covered jaws
$41.40 - $72.80
Specifications
Type Extension Clamp
Includes Fiberglass and Vinyl sleeve
View More Specs
Includes: Vinyl and fiberglass sleeves, extension arm O.D.: 0.43 in. (1.1cm)
Products 2
Catalog Number Length (English) Length (Metric) Size (English) Grip Size (Metric) Grip Price Quantity & Availability  
Catalog Number Length (English) Length (Metric) Size (English) Grip Size (Metric) Grip Price Quantity & Availability  
05-769-3Q
View Documents
9 in. 22.86 cm 2.5 in. 6.35 cm
Each for $72.80
Add to cart
05-769-4Q
View Documents
8 in. 20.32 cm 1.5 in. 3.81 cm
Each for $41.40
Add to cart
Description
Description
Flat, vinyl- or fiberglass-covered jaws
Hold beakers and flasks firmly at any angle, variable distance from support.
  • One jaw is fixed, other opens against a stainless-steel compression spring
  • Opening is gradual to maximum angle, without binding
  • Noncorrosive
  • Easy-to-clean clamps are nickel-plated zinc with seamless brass extension tube, brass compressing screw, and pivot pin
  • Without clamp holder
Castaloy clamps and holders should not be subjected to direct flame or other intense heat sources. At continuous temperatures in excess of 150°C (302°F), some significant distortion can occur. Fiberglass sleeves are recommended for all applications above 100°C (212°F).
Specifications
Specifications
Extension Clamp
Fiberglass and Vinyl sleeve