Learn More
Fisherbrand™ Castaloy™ VariFlow™ Pinchcock
Flowrates duplicable time after time
Catalog No. 05-849-5Q
Add to cart
Description
Description
  • With thumbscrew and locking nut for selecting degree of closure
  • Use effectively on up to 0.75 in. (9.5mm) O.D. flexible tubing
  • Does not cut tubing, rust, or corrode
Specifications
Specifications
VariFlow Pinchcock
Thumbscrew and locking nut