Learn More
Thermo Scientific™ Chemically-Preserved Environmental Sample Containers
Provide an extra measure of assurance that proper sampling protocols are followed
Manufacturer:  Thermo Scientific™ PP14040CEP.5ST
Catalog No. 12-101-905
Add to cart
Includes: Certified sample containers dosed with chemical preservative; certificate of analysis for container and preservative, individually barcoded containers traceable to certs, sample analysis labels unattached for optional use, hazardous material compliant packaging. MSDS available upon request.
Description
Description
  • Standard Thermo Scientific Certified sample containers chemically-preserved with certified chemical preservatives for environmental sample analyses
  • Packaged in compliance with the shipping requirements of 49 CFR Part 173.4 DOT Small Quantities Classification for highway and rail transport
Custom preserved containers available by special order:
  • Choose a standard certified Thermo Scientific environmental sample container
  • Choose a chemical from our standard chemical list
  • Specify the volume and concentration of chemical to aliquot into each container
  • 10 Case minimum order
Order Info
Products available only in the US
Specifications
Specifications
Clear
40 mL clear septa vial w/0.5 mL sodium thiosulfate
24-414
Environmental Sampling
Packaged in compliance with the shipping requirements of 49 CFR Part 173.4 DOT Small Quantities Classification for highway and rail transport