Thermo Scientific™ CNET 1500 NDS™ Network Docking Station Available on GSA/VA Contract for Federal Government customers only.
An easily-implemented, reliable, and self-contained real time wireless radiation and environmental monitoring solution
Manufacturer: Thermo Scientific™ SYS/1/70725/100
 View more versions of this product
Catalog No. 19-166-745
  Check Availability
Add to Cart
Required Accessories : AC-power
    Specifications
    For Use With (Application) Air quality monitoring in nuclear power, security and plant operations, radiography coverage and monitoring, decommissioning and reclamation
    Product Type Network Docking Station
    A versitle device that is both an Area Radiation Monitor and a hub for connecting radiation and environmental monitoring devices to a wireless network
    • Minimal setup and maintenance minimizes personnel exposure time
    • Uses proven Thermo technologies: EPD, CNET 1500, and wireless network
    • Easily communicates with base station using Thermo Scientific Mk2 Radio
    • Standard AC mains power with battery backup
    • Integral light and sound alarms driven by embedded intelligent controller
    • Front panel LEDs verify power, probe, and radio operation
    • Power on self test verifies intelligent controller and component operation
    • Uses industry standard 2.4 GHz wireless communications with extended wireless range (up to five miles) using high-gain antennas and/or repeaters
    • Easy to integrate with existing systems such as Thermo Scientific ViewPoint remote monitoring and data management system
    • Rugged portable enclosure
    • RadReachBack™ capability with optional PDT-100 Satellite transceiver
    • Useful for health physicists, plant workers, security teams, and first responders
      • Air quality monitoring in nuclear power, security, and plant operations
      • Radiography coverage and monitoring
      • Decommissioning and reclamation
      • Public event security such as concerts, sporting events, and high profile gatherings
      • Hazmat teams and first-responder accident coverage in buildings, subways, and many other locations allowing simple communications with base station
  • Description & Specifications
Related Products
Product Content Feedback