Learn More
Epredia™ Permafrost™ Slides
Available in a wide variety of colors to ease identification.
Supplier: Epredia™ 4443
This item is not returnable.
View return policy
Description
- Resistant to commonly used laboratory solvents
- 1.0 to 1.2 mm thick with ground edges
Specifications
Permafrost 1 End | |
1 to 1.2 mm | |
Red |
Glass | |
Ground | |
1440/Cs. |