Learn More
Epredia™ Colormark™ Slide Cartridges forSHUR/Mark™ Labeling Systems
Background color makes etched patient information more visible
Supplier: Epredia™ CM6951W001
This item is not returnable.
View return policy
Description
- Brightly coated with contrasting background color
- Self-contained packages ready for loading into your SHUR/Mark slide labeling system
- Simply remove the cover from the slide package and drop the sleeve into the loading bin of the SHUR/Mark labeling system
- Available with Plus formula to provide the utmost adhesion
Specifications
Green | |
Non-adhesion | |
144 Pk., 10 Pk./Cs. |