Learn More
Epredia Colormark and Colormark Plus Slides
Background color makes etched patient information more visible.
$94.70 - $915.50
Catalog Number | Mfr. No. | Color | Coating | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Coating | Price | Quantity & Availability | ||||
22-230-904
![]() |
Epredia
CM9951WA001 |
Aqua | Non-Adhesion |
Case of 1440 for $915.50
|
|
22-038-330
|
Epredia
CM9951WAPLUS001 |
Aqua | Adhesion |
Pack of 144 for $94.70
Case of 10 PK for $938.00 |
|
22-034-986
![]() |
Epredia
CM9951WBOPS001 |
Burnt Orange | Adhesion |
Pack of 144 for $126.70
Case of 10 PK for $1,088.00 |
|
22-230-902
|
Epredia
CM9951BO001 |
Burnt Orange | Non-Adhesion |
Pack of 144 for $142.60
Case of 10 PK for $1,290.00 |
|
22-038-332
|
Epredia
CM9951WGLPS001 |
Gold | Adhesion |
Pack of 144 for $94.70
Case of 10 PK for $938.00 |
|
22-230-903
![]() |
Epredia
CM9951WT001 |
Tan | Non-Adhesion |
Pack of 144 for $142.60
Case of 10 PK for $1,290.00 |
|
22-038-331
|
Epredia
CM9951WTPLUS001 |
Tan | Adhesion |
Pack of 144 for $94.70
Case of 10 PK for $938.00 |
|
22-038329
|
Epredia
CM9951WL001 |
Lavender | Non-Adhesion |
Pack of 144 for $142.60
Case of 10 PK for $1,290.00 |
|
22-038328
![]() |
Epredia
CM9951WLPLUS001 |
Lilac/Black | Adhesion |
Pack of 144 for $215.50
Case of 10 PK for $1,815.00 |
|
22-035814
|
Epredia
CM4951W001 |
White | Non-Adhesion |
Pack of 144 for $123.30
Case of 10 PK for $1,059.00 |
|
22-038324
![]() |
Epredia
CM4951WPLUS001 |
White | Adhesion |
Pack of 144 for $221.00
Case of 10 PK for $1,867.00 |
|
22-035815
|
Epredia
CM5951W001 |
Blue | Non-Adhesion |
Pack of 144 for $123.70
Case of 10 PK for $1,062.00 |
|
22-038325
|
Epredia
CM5951WPLUS001 |
Blue | Adhesion |
Pack of 144 for $221.50
Case of 10 PK for $1,866.00 |
|
22-035816
|
Epredia
CM6951W001 |
Green | Non-Adhesion |
Pack of 144 for $123.30
Case of 10 PK for $1,059.00 |
|
22-038326
|
Epredia
CM6951WPLUS001 |
Green | Adhesion |
Pack of 144 for $221.00
Case of 10 PK for $1,866.00 |
|
22-035817
|
Epredia
CM7951W001 |
Pink | Non-Adhesion |
Pack of 144 for $123.80
Case of 10 PK for $1,064.00 |
|
22-038323
|
Epredia
CM7951WPLUS001 |
Pink | Adhesion |
Pack of 144 for $221.00
Case of 10 PK for $1,868.00 |
|
22-035818
|
Epredia
CM8951W001 |
Yellow | Non-Adhesion |
Pack of 144 for $123.50
Case of 10 PK for $1,061.00 |
|
22-038327
|
Epredia
CM8951WPLUS001 |
Yellow | Adhesion |
Pack of 144 for $221.00
Case of 10 PK for $1,866.00 |
|
Description
- Brightly coated with contrasting background color
- Simple to use self-contained packages ready for loading into etching slide printers
- Available in adhesion and non-adhesion varieties
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.