Learn More
Thermo Scientific™ Crystal Plates and Spare Parts for the Thermo Scientific™ iD7 ATR Accessory
Choose the crystal plate to meet specific application and budget requirements
Manufacturer:  Thermo Scientific™ 869169100
 View more versions of this product
Catalog No. 14-387-804
Add to cart
Description
Description
Configure the iD7 ATR accessory with one or more of the crystal plates:
  • Diamond (AR-coated or uncoated) for liquids and all solids
  • ZnSe for liquids and soft solids
  • Germanium for highly absorbing materials like black rubbers
Spare part kit includes iD7 ATR tip and volatiles cover.
Specifications
Specifications
iD7 Germanium Plate