Learn More
Thermo Scientific™ D.A.R.T.'S™ (Disposable Automation Research Tips)
D.A.R.T.'S (Disposable Automation Research Tips) have been designed for the ultimate performance in automated pipetting
Manufacturer:  Thermo Scientific™ 551611
 View more versions of this product
Catalog No. 14-387-677
Add to cart
Includes: 20 magazines per case
Description
Description
Unlike conventional automation tips, D.A.R.T. tips are provided in disposable rigid magazines that seal onto the pipetting heads by sealing against a silicone pad. This forms a definitive seal which minimizes tip-to-tip height variations and elimiates static interactions, enabling consistent dispensing into the bottom of microplate wells.
  • Compatible with the Matrix Hydra DT and Matrix PlateMate 2x3 and PlateMate Plus Systems
  • Stepped-body design minimizes volume displacement and ensures tip straightness for better well access
  • Highly polished molds and cores ensure low liquid retention
  • Small internal tip orifice provides better small volume droplet formation, resulting in optimal accuracy and precision
  • Sterile, sterile filtered, wide-bore, and extended-length tip options available, offering the flexibility to choose tips for any application
  • Thorough inspection for proper formation as well as straightness ensures tips are free of defects
  • Unique face seal design ensures consistent tip-to-tip height for precise low volume dispensing
  • Entire assembly can be quickly removed and replaced, eliminating extra handling of the tips which can cause damage or contamination
Specifications
Specifications
Automation, Extended Length
300μL
Thermo Scientific™ Matrix Hydra DT
Thermo Scientific™ Matrix Hydra DT, Thermo Scientific™ Matrix PlateMate 2x3, Thermo Scientific™ Versette™
Sterile