Learn More
Thermo Scientific™ EPD Mk2, G and N2 Readers
To complement the Thermo Scientific EP DMk2, G and N2. The basic IrDA reader, is ideal for office environments and mobile applications
Supplier: Thermo Scientific™ EPD/1/62570/000
Description
Compatible with the full range of Mk2 software products
- Connect directly to the PC via a serial or optional USB interface
- Require no power supply
- For more demanding environments, and where it is necessary to ensure that only one specific EPD can be “seen” by the reader, the desktop model ACT-5 may be used
- The IrDA and ACT-5 are compatible with EasyEPD2 and EasyIssue software
Specifications
0.1 lb. | |
Serial RS232 or USB 1.1 and 2.0 compatible interface direct to PC | |
USB interface | |
1 m |
50 g | |
For office Environments and Mobile applications | |
3.2 ft. | |
Basic Irda Reader |