Learn More
Thermo Scientific™ Evolution™ 300 and 600 Spectrophotometer Solid Sample Holders
Designed to hold solid samples for transmittance measurements
Manufacturer:  Thermo Scientific™ 222217400
 View more versions of this product
Catalog No. 14-387-504
Add to cart
Description
Description
Solid Sample Holder
  • For use with Thermo Scientific™ Evolution™ 300 and 600 Spectrophotometers
  • Provides easy mounting of solid samples like filters, plastic, glass and optics
  • Accommodates standard 2 x 3 in. sample holders
Universal Sample Holder
  • Holds round or square optical samples; glass or plastic samples; lenses from sunglasses or goggles
  • Requires one Solid Sample Holder (14-387-503)
Specifications
Specifications
Thermo Scientific™ Evolution™ 300 and 600 UV-visible spectrophotometer
Round or square optical samples, glass or Plastic samples, lenses from sunglasses or goggles
Universal Sample Holder