Thermo Scientific™ Evolution™ 60S UV-Visible Spectrophotometer
From the technician to the research scientist, the Evolution 60S offers the ideal solution delivering power and flexibility in a compact package
Efficient optical design delivers a new level of clarity to your measurements.
$5922.82 - $6675.97
Specifications
Detector Type Dual Silicon Photodiodes
Resolution >1.5
Connection Type USB
Scan Speed 4200nm/min
Wavelength Range 190 to 1100nm
View More Specs
  • Products 3
  • Description & Specifications
Products
Catalog Number Mfr. No. Product Type Price Quantity & Availability    
14-386-510
Accessories
Thermo Scientific
840-208500
Evolution 60S w/local control Each for $6,175.57
Check Availability
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
14-386-511
Accessories
Thermo Scientific
840-208600
Evolution 60S w/local control and printer Each for $6,675.97
Check Availability
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
14-386-512
Accessories
Thermo Scientific
840-208700
Evolution 60S w/PC control Each for $5,922.82
Check Availability
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Descriptions & Specifications
Related Products