Learn More
Thermo Scientific™ F-G1 Fixed Angle Adapters For CL10™ Rotor
For Thermo Scientific™ IEC F-G1 Fixed Angle rotor
Specifications
Type Adapter
For Use With (Equipment) Thermo Scientific™ IEC™ F-G1 fixed angle rotor
View More Specs
Products 4
Catalog Number Mfr. No. Capacity Max. Tube Diameter Max. Tube Length Price Quantity & Availability  
Catalog Number Mfr. No. Capacity Max. Tube Diameter Max. Tube Length Price Quantity & Availability  
05-376-238
View Documents
Thermo Scientific™
11210527
1 x 16/15 mL 18 mm 107 mm Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. N/A
05-376-239
View Documents
Thermo Scientific™
11210532
1 x 15 mL 16.5 mm 121 mm Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. N/A
05-376-240
View Documents
Thermo Scientific™
11210537
1 x 10 mL 16 mm 82 mm Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. N/A
05-376-241
View Documents
Thermo Scientific™
11210542
1 x 2/1.5 mL 10 mm 41 mm Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. N/A
Description
Description
For CL10 rotor. Sold in a set of 6.
Specifications
Specifications
Adapter
Thermo Scientific™ IEC™ F-G1 fixed angle rotor