Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Buffers for GeneJET™ Blood RNA Kit

Purify high quality total RNA from fresh or frozen whole blood, as well as blood collected in PAXgene and Tempus Tubes

Manufacturer:  Thermo Scientific™ R2841

 View more versions of this product

Catalog No. FERR2841


Add to cart

Description

Description

The GeneJET™ Stabilized and Fresh Whole Blood RNA Kit is designed for rapid and efficient purification of high quality total RNA from various whole blood samples: fresh, frozen, or human blood samples stabilized with commercial stabilizers such as Tempus™ and PAXgene™.
The kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation. The kit includes a DNase l for the enzymatic removal of genomic DNA. RNA can be isolated from whole blood in less than 30 minutes (including cell lysis and DNase l treatment).
Highlights
·Versatile - compatible with fresh and frozen blood, buffy coat, as well as blood collected in PAXgene™and Tempus ™ Tubes
·Convenient - specially optimized Lysis Buffer is suitable for RNA stabilization up to 48 h
·Efficient - up to 4 µg of total RNA from 500 µL of healthy human blood
·Pure - isolates intact total RNA free of any RT-qPCR inhibitors
Applications
Isolated total RNA can be used in:
· RT-PCR
· RT-qPCR
· Northern Blotting
Includes
· Lysis Buffer
· Wash Buffer 1 (conc.)
· Wash Buffer 2 (conc.)
· DNase l (lyophilized)
· DNase I Reconstitution Buffer
· 2x DNase I Buffer
· Manganese Chloride Solution
· Water, nuclease-free
· GeneJET RNA Purification Columns pre-assembled with Collection Tubes
· Collection Tubes, 2 mL
· Collection Tubes, 1.5 mL

Hazardous:No
> Quality Control:Thermo Scientific GeneJET Stabilized and Fresh Whole Blood RNA Purification Mini Kit is qualified by isolating total RNA from 250 µL of human whole blood following the protocol outlined in the manual. The quality of isolated RNA is evaluated spectrophotometrically and by agarose gel electrophoresis. The purified RNA has an A260/A280 ratio between 1.9 and 2.1 and the RNA integrity number (RIN) of ≥ 8.
> Shelf Life:24 Months
> Shipping Condition:Ambient
> Storage Condition:Ambient

Specifications

Specifications

GeneJET Blood RNA Kit
40mL
Store at room temperature
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit