Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ClaSeek™ Library Preparation Kit, Ion Torrent™ Compatible

Generate Illumina™-compatible next-gen sequencing libraries from low sample input using a one-tube PCR-free method with optimized master mixes

Manufacturer:  Thermo Scientific™ K1351

 View more versions of this product

Catalog No. FERK1351

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Thermo Scientific ClaSeek Library Preparation Kit, Illumina™ compatible, is designed for fast and convenient construction of PCR-free NGS fragment library from DNA sample input as low as 500 ng. In combination with the amplification protocol the kit also enables the generation of DNA fragment libraries from ultra-low 5 ng sample input.
The kit utilizes a highly efficient NGS-optimized library construction protocol combining end-conversion and adapter addition steps into a simple and convenient one-tube procedure. This minimizes unnecessary pipetting steps and reduces hands-on time, allowing for PCR-free library construction in less than 70 minutes. The kit is suitable for construction of 50, 300, 500 bp read-length Illumina™-compatible libraries and is validated for use on Illumina™ HiSeq™ and MiSeq™ sequencing platforms.
Highlights
·PCR-free - significantly minimized bias and improved coverage with amplification-free protocol
·Flexible - universal protocol for variable sample input (5 ng - 1 µg) and DNA fragment lengths
·Fast and Convenient - fewer cleanup steps, less hands-on time, optimized master mixes, one-tube end conversion procedure.
Applications
DNA fragment library preparation for downstream use in next generation sequencing workflow.
Includes
· ClaSeek End Conversion Master Mix, IL
· ClaSeek Ligation Mix, IL
· Water, nuclease-free

Specifications

Specifications

ClaSeek End Conversion Master Mix IT, ClaSeek Ligation Mix IT, Adapters IT, Library Amplification Primer Mix, nuclease-free water, Cleanup Columns (pre-assembled with collection tubes)
DNA fragment library preparation for downstream use in next generation sequencing workflow.
10 reactions
Store at -20deg.C. Cleanup Columns must be stored at room temperature.
Safety and Handling

Safety and Handling

ShelfLife : 1 Year

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit